Assay Details
| Gene Symbol: Sgpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009163.3 | NP_033189.2 | 13 - 14 | 1634 | 129 | |
| GenBank mRNA | AF036894.1 | - | 12 - 13 | 1504 | 129 | |
| AK036747.1 | - | 13 - 14 | 1522 | 129 | ||
| AK037789.1 | - | 13 - 14 | 1519 | 129 | ||
| AK043024.1 | - | 13 - 14 | 1521 | 129 | ||
| AK049342.1 | - | 13 - 14 | 1542 | 129 | ||
| AK075851.1 | - | 4 - 5 | 619 | 129 | ||
| AK150053.1 | - | 13 - 14 | 1529 | 129 | ||
| AK173137.1 | - | 13 - 14 | 1563 | 129 | ||
| BC026135.1 | - | 13 - 14 | 1580 | 4212293 | 129 | |
| CT010214.1 | - | 12 - 13 | 1444 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sgpl1
Genomic Map