Assay Details
| Gene Symbol: Sgpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009163.3 | NP_033189.2 | 4 - 5 | 453 | 143 | |
| GenBank mRNA | AF036894.1 | - | 3 - 4 | 323 | 143 | |
| AK036747.1 | - | 4 - 5 | 341 | 143 | ||
| AK037789.1 | - | 4 - 5 | 338 | 143 | ||
| AK043024.1 | - | 4 - 5 | 340 | 143 | ||
| AK049342.1 | - | 4 - 5 | 361 | 143 | ||
| AK150053.1 | - | 4 - 5 | 348 | 143 | ||
| AK173137.1 | - | 4 - 5 | 382 | 143 | ||
| BC026135.1 | - | 4 - 5 | 399 | 4212293 | 143 | |
| CT010214.1 | - | 3 - 4 | 263 | 143 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sgpl1
Genomic Map