Assay Details
| Gene Symbol: Susd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162913.1 | NP_001156385.1 | 3 - 4 | 485 | 69 | |
| NM_027890.5 | NP_082166.3 | 5 - 6 | 845 | 69 | ||
| GenBank mRNA | AK004703.2 | - | 5 - 6 | 815 | 69 | |
| AK028817.1 | - | 5 - 6 | 821 | 69 | ||
| AK033823.1 | - | 3 - 4 | 458 | 69 | ||
| AK037059.1 | - | 5 - 6 | 827 | 69 | ||
| AK037127.1 | - | 5 - 6 | 796 | 69 | ||
| AK038944.1 | - | 5 - 6 | 846 | 69 | ||
| AK042469.1 | - | 5 - 6 | 829 | 69 | ||
| AK128959.1 | - | 5 - 6 | 817 | 69 | ||
| AK167155.1 | - | 5 - 6 | 846 | 69 | ||
| BC026837.1 | - | 5 - 6 | 798 | 4222578 | 69 | |
| BC066833.1 | - | 3 - 4 | 463 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK038944.1, AK167155.1 transcripts.
Set Membership: |
|

Susd2
Genomic Map