Assay Details
| Gene Symbol: Susd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162913.1 | NP_001156385.1 | 6 - 7 | 1034 | 77 | |
| NM_027890.5 | NP_082166.3 | 8 - 9 | 1394 | 77 | ||
| GenBank mRNA | AK004703.2 | - | 8 - 9 | 1364 | 77 | |
| AK028817.1 | - | 8 - 9 | 1370 | 77 | ||
| AK033823.1 | - | 6 - 7 | 1007 | 77 | ||
| AK037059.1 | - | 8 - 9 | 1376 | 77 | ||
| AK037127.1 | - | 8 - 9 | 1345 | 77 | ||
| AK038944.1 | - | 6 - 7 | 1662 | 77 | ||
| AK042469.1 | - | 8 - 9 | 1378 | 77 | ||
| AK128959.1 | - | 8 - 9 | 1366 | 77 | ||
| AK167155.1 | - | 7 - 8 | 1576 | 77 | ||
| BC026837.1 | - | 8 - 9 | 1347 | 4222578 | 77 | |
| BC066833.1 | - | 5 - 6 | 1193 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Susd2
Genomic Map