Assay Details
| Gene Symbol: Sar1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009120.2 | NP_033146.1 | 2 - 3 | 166 | 88 | |
| GenBank mRNA | AK157843.1 | - | 2 - 3 | 178 | 88 | |
| AK159239.1 | - | 2 - 3 | 177 | 88 | ||
| AK168175.1 | - | 2 - 3 | 178 | 88 | ||
| BC005549.1 | - | 2 - 3 | 167 | 3585667 | 88 | |
| L20294.1 | - | 2 - 3 | 141 | 88 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK129305.1 | - | 3364 | Chr. - - [-] on Build | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sar1a
Genomic Map