Assay Details
| Gene Symbol: Cgnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB186125.1 | - | 7 - 8 | 2725 | 81 | |
| BB667316.1 | - | 2 - 3 | 212 | 81 | ||
| BC139006.1 | - | 6 - 7 | 2197 | 8862028 | 81 | |
| BC158061.1 | - | 6 - 7 | 2200 | 9007361 | 81 | |
More Information
Set Membership: |
|

Cgnl1
Genomic Map