Assay Details
| Gene Symbol: Ppan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145610.2 | NP_663585.2 | 7 - 8 | 818 | 65 | |
| GenBank mRNA | AK039027.1 | - | 7 - 8 | 723 | 65 | |
| AK171672.1 | - | 3 - 4 | 484 | 65 | ||
| AK172615.1 | - | 3 - 4 | 484 | 65 | ||
| BC014688.1 | - | 7 - 8 | 736 | 4489442 | 65 | |
| BC059277.1 | - | 6 - 7 | 902 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppan
Genomic Map