| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900015347.mm.1 |
NM_001082485.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015347.mm.1 |
NM_001082485.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015347.mm.1 |
NM_001135019.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015347.mm.1 |
NM_001135019.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015347.mm.1 |
TC0900001838.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900015348.mm.1 |
NM_001082485.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015348.mm.1 |
NM_001082485.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015348.mm.1 |
NM_001135019.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015348.mm.1 |
NM_001135019.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015348.mm.1 |
TC0900001838.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900015349.mm.1 |
NM_001082485.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015349.mm.1 |
NM_001082485.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015349.mm.1 |
NM_001135019.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015349.mm.1 |
NM_001135019.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015349.mm.1 |
TC0900001838.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900015350.mm.1 |
NM_001082485.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015350.mm.1 |
NM_001082485.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015350.mm.1 |
NM_001135019.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015350.mm.1 |
NM_001135019.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015350.mm.1 |
TC0900001838.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900015352.mm.1 |
NM_001082485.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015352.mm.1 |
NM_001082485.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015352.mm.1 |
NM_001135019.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015352.mm.1 |
NM_001135019.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015352.mm.1 |
TC0900001838.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900015346.mm.1 |
NM_001082485.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015346.mm.1 |
NM_001082485.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015346.mm.1 |
NM_001135019.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015346.mm.1 |
NM_001135019.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015346.mm.1 |
TC0900001838.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900015351.mm.1 |
NM_001082485.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015351.mm.1 |
NM_001082485.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015351.mm.1 |
NM_001135019.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015351.mm.1 |
NM_001135019.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900015351.mm.1 |
TC0900001838.mm.1 |
MTA TC |