Assay Details
| Gene Symbol: Zfp266 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082485.1 | NP_001075954.1 | 8 - 9 | 877 | 136 | |
| NM_001135019.1 | NP_001128491.1 | 8 - 9 | 866 | 136 | ||
| GenBank mRNA | BC048701.1 | - | 8 - 9 | 817 | 136 | |
| BC061209.1 | - | 8 - 9 | 863 | 136 | ||
| BC089552.1 | - | 8 - 9 | 876 | 136 | ||
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P360880 | BM941296.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P360880 | AK088432.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P360880 | NM_001082485.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P360880 | NM_001135019.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P360880 | BM941296.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P360880 | AK088432.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P360880 | NM_001082485.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P360880 | NM_001135019.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2174816 | BC089552.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2174816 | BC048701.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2174816 | BC061209.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2174816 | NM_001082485.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2174816 | NM_001135019.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 107246 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 143700 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 567557 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 15666 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 186847 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 832237 | AK033992.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 832237 | BC104120.2 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 832237 | NM_147049.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 444746 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 855818 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 245222 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 77861 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181456 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 874089 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 490931 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 81163 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 581686 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 541311 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 97046 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 150050 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 102138 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 81957 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128214 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113826 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2174816 | BC089552.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2174816 | BC048701.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2174816 | BC061209.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2174816 | NM_001082485.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2174816 | NM_001135019.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P360880 | BM941296.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P360880 | AK088432.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P360880 | NM_001082485.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P360880 | NM_001135019.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P360880 | BM941296.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P360880 | AK088432.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P360880 | NM_001082485.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P360880 | NM_001135019.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113826 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06716213 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128214 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01880613 | - | - |
| Whole Mouse Genome G4122A | A_52_P312084 | AK081787.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P360880 | BM941296.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P360880 | AK088432.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P360880 | NM_001082485.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P360880 | NM_001135019.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P360880 | BM941296.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P360880 | AK088432.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P360880 | NM_001082485.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P360880 | NM_001135019.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P722599 | NM_001082485.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P722599 | NM_001135019.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P722599 | AK020507.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103984 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128214 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06716213 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113826 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07906263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07642606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07643686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342798 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26342785 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10591259 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591255 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591254 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591256 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591262 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591257 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591261 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591258 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591260 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434084_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1431206_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1457913_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454812_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10591253 | - | - |
| Mouse Genome 430 2.0 Array | 1441596_at | - | - |
| Mouse Genome 430 2.0 Array | 1441596_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431206_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1457913_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434084_at | - | - |
| Mouse Genome 430 2.0 Array | 1454812_at | - | - |
| Mouse Genome 430 2.0 Array | 1431206_at | - | - |
| Mouse Genome 430 2.0 Array | 1454812_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434084_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1457913_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 77519_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 77519_PM_at | - | - |
| Mouse Expression Set 430 | 1457913_PM_at | - | - |
| Mouse Expression Set 430 | 1434084_at | - | - |
| Mouse Expression Set 430 | 1454812_PM_at | - | - |
| Mouse Expression Set 430 | 1457913_at | - | - |
| Mouse Expression Set 430 | 1454812_at | - | - |
| Mouse Expression Set 430 | 1434084_PM_at | - | - |
| Mouse Expression Set 430 | 1431206_at | - | - |
| Mouse Expression Set 430 | 1431206_PM_at | - | - |
| Murine Genome U74v2 Set | 166198_f_at | - | - |
| Murine Genome U74v2 Set | 166198_PM_f_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454812_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434084_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431206_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457913_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457913_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454812_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1434084_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431206_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 382735 | - | - |
| ABI Mouse Genome Survey Microarray | 908960 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223473 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1222946 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1243686 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2610683 | - | - |

Zfp266
Genomic Map