Assay Details
| Gene Symbol: Ccpg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114328.2 | NP_001107800.1 | 3 - 4 | 363 | 63 | |
| NM_001286544.1 | NP_001273473.1 | 3 - 4 | 363 | 63 | ||
| NM_001286545.1 | NP_001273474.1 | 3 - 4 | 363 | 63 | ||
| NM_001286546.1 | NP_001273475.1 | 3 - 4 | 363 | 63 | ||
| NM_028181.5 | NP_082457.3 | 3 - 4 | 363 | 63 | ||
| GenBank mRNA | AK007973.1 | - | 3 - 4 | 260 | 63 | |
| AK032645.1 | - | 3 - 4 | 249 | 63 | ||
| AK076325.2 | - | 3 - 4 | 290 | 63 | ||
| AK128894.1 | - | 3 - 4 | 289 | 63 | ||
| AK134675.1 | - | 3 - 4 | 260 | 63 | ||
| BC043049.1 | - | 3 - 4 | 262 | 5704185 | 63 | |
| BC082602.1 | - | 3 - 4 | 264 | 30663407 | 63 | |
| CN721780.1 | - | 4 - 5 | 476 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001286546.1 transcripts.
Set Membership: |
|

Ccpg1
Genomic Map