Assay Details
| Gene Symbol: B3gat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB055781.1 | - | 2 - 3 | 348 | 81 | |
| AK041316.1 | - | 2 - 3 | 420 | 81 | ||
| AK044599.1 | - | 447 | 81 | |||
| AK082739.1 | - | 2 - 3 | 463 | 81 | ||
| AK220561.1 | - | 2 - 3 | 455 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB055781.1, AK041316.1, AK044599.1, AK082739.1 transcripts.
Set Membership: |
|

B3gat1
Genomic Map