Assay Details
Gene Symbol: Tbcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025548.3 | NP_079824.2 | 2 - 3 | 592 | 97 | |
GenBank mRNA | AK002316.1 | - | 2 - 3 | 333 | 97 | |
AK003655.2 | - | 2 - 3 | 338 | 97 | ||
BC010684.1 | - | 2 - 3 | 325 | 2648112 | 97 | |
BQ445322.1 | - | 2 - 3 | 592 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
