Assay Details
Gene Symbol: Dhx32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286030.1 | NP_001272959.1 | 5 - 6 | 1576 | 81 | |
NM_001286031.1 | NP_001272960.1 | 5 - 6 | 1445 | 81 | ||
NM_001286032.1 | NP_001272961.1 | 4 - 5 | 1015 | 81 | ||
NM_133941.2 | NP_598702.1 | 5 - 6 | 1529 | 81 | ||
GenBank mRNA | AK028910.1 | - | 5 - 6 | 1386 | 81 | |
AK033624.1 | - | 5 - 6 | 1531 | 81 | ||
AK079184.1 | - | 4 - 5 | 915 | 81 | ||
AK135760.1 | - | 5 - 6 | 1578 | 81 | ||
AK167889.1 | - | 5 - 6 | 1474 | 81 | ||
AK169005.1 | - | 5 - 6 | 1520 | 81 | ||
AK172504.1 | - | 5 - 6 | 1510 | 81 | ||
AK177046.1 | - | 1 - 2 | 67 | 81 | ||
AY065980.1 | - | 5 - 6 | 1531 | 81 | ||
BC006911.1 | - | 4 - 5 | 1086 | 81 | ||
BC022920.1 | - | 4 - 5 | 1285 | 4460064 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
