Assay Details
Gene Symbol: Dhx32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286030.1 | NP_001272959.1 | 8 - 9 | 2031 | 74 | |
NM_001286031.1 | NP_001272960.1 | 8 - 9 | 1900 | 74 | ||
NM_001286032.1 | NP_001272961.1 | 7 - 8 | 1470 | 74 | ||
NM_133941.2 | NP_598702.1 | 8 - 9 | 1984 | 74 | ||
GenBank mRNA | AK028910.1 | - | 8 - 9 | 1841 | 74 | |
AK033624.1 | - | 8 - 9 | 1986 | 74 | ||
AK079184.1 | - | 7 - 8 | 1370 | 74 | ||
AK135760.1 | - | 8 - 9 | 2034 | 74 | ||
AK167889.1 | - | 8 - 9 | 1929 | 74 | ||
AK169005.1 | - | 8 - 9 | 1975 | 74 | ||
AK172504.1 | - | 8 - 9 | 1965 | 74 | ||
AY065980.1 | - | 8 - 9 | 1986 | 74 | ||
BC006911.1 | - | 7 - 8 | 1541 | 74 | ||
BC022920.1 | - | 7 - 8 | 1740 | 4460064 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
