Assay Details
| Gene Symbol: Stard3nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK030365.1 | - | 1 - 2 | 99 | 87 | |
| AK152116.1 | - | 1 - 2 | 99 | 87 | ||
| BG084215.2 | - | 1 - 2 | 55 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK152116.1 transcripts.
Set Membership: |
|

Stard3nl
Genomic Map