Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600000060.mm.1 |
TC0600000004.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600000060.mm.1 |
NM_001167750.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000060.mm.1 |
NM_001167750.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000060.mm.1 |
NM_024260.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000060.mm.1 |
NM_024260.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000062.mm.1 |
TC0600000004.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600000062.mm.1 |
NM_001167750.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000062.mm.1 |
NM_001167750.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000062.mm.1 |
NM_024260.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000062.mm.1 |
NM_024260.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000063.mm.1 |
TC0600000004.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600000063.mm.1 |
NM_001167750.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000063.mm.1 |
NM_001167750.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000063.mm.1 |
NM_024260.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000063.mm.1 |
NM_024260.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000061.mm.1 |
TC0600000004.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600000061.mm.1 |
NM_001167750.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000061.mm.1 |
NM_001167750.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000061.mm.1 |
NM_024260.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000061.mm.1 |
NM_024260.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000057.mm.1 |
TC0600000004.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600000057.mm.1 |
NM_001167750.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000057.mm.1 |
NM_001167750.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000057.mm.1 |
NM_001167751.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000057.mm.1 |
NM_001167751.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000057.mm.1 |
NM_024260.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000057.mm.1 |
NM_024260.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000065.mm.1 |
TC0600000004.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600000065.mm.1 |
NM_001167750.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000065.mm.1 |
NM_001167750.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000065.mm.1 |
NM_024260.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000065.mm.1 |
NM_024260.5 |
Refseq |