Assay Details
| Gene Symbol: Ccdc132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167750.1 | NP_001161222.1 | 20 - 21 | 1986 | 85 | |
| NM_001167751.1 | NP_001161223.1 | 20 - 21 | 1986 | 85 | ||
| NM_024260.5 | NP_077222.4 | 20 - 21 | 1986 | 85 | ||
| GenBank mRNA | AK006602.1 | - | 2 - 3 | 212 | 85 | |
| AK042031.1 | - | 20 - 21 | 1966 | 85 | ||
| AK075898.1 | - | 20 - 21 | 1975 | 85 | ||
| AK129458.1 | - | 20 - 21 | 1936 | 85 | ||
| AK134533.1 | - | 4 - 5 | 3785 | 85 | ||
| BC002304.1 | - | 5 - 6 | 531 | 3591600 | 85 | |
| BC036294.1 | - | 20 - 21 | 1926 | 3500316 | 85 | |
| BC043092.1 | - | 20 - 21 | 1943 | 6405356 | 85 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK006602.1 transcripts.
Set Membership: |
|

Ccdc132
Genomic Map