Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600000049.mm.1 |
TC0600000004.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600000049.mm.1 |
NM_001167750.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000049.mm.1 |
NM_001167750.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000049.mm.1 |
NM_001167751.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000049.mm.1 |
NM_001167751.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000049.mm.1 |
NM_024260.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000049.mm.1 |
NM_024260.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000048.mm.1 |
TC0600000004.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600000048.mm.1 |
NM_001167750.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000048.mm.1 |
NM_001167750.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000048.mm.1 |
NM_001167751.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000048.mm.1 |
NM_001167751.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000048.mm.1 |
NM_024260.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000048.mm.1 |
NM_024260.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000050.mm.1 |
TC0600000004.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600000050.mm.1 |
NM_001167750.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000050.mm.1 |
NM_001167750.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000050.mm.1 |
NM_001167751.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000050.mm.1 |
NM_001167751.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000050.mm.1 |
NM_024260.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600000050.mm.1 |
NM_024260.5 |
Refseq |