Assay Details
| Gene Symbol: Wnt8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009290.2 | NP_033316.1 | 1 - 2 | 110 | 93 | |
| GenBank mRNA | BC119212.1 | - | 1 - 2 | 110 | 8733961 | 93 |
| BC120517.1 | - | 1 - 2 | 110 | 40129440 | 93 | |
| Z68889.1 | - | 1 - 2 | 125 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wnt8a
Genomic Map