Assay Details
| Gene Symbol: Dsc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013505.3 | NP_038533.1 | 12 - 13 | 2141 | 71 | |
| GenBank mRNA | AK028604.1 | - | 13 - 14 | 2181 | 71 | |
| AK030333.1 | - | 12 - 13 | 2142 | 71 | ||
| AK048547.1 | - | 12 - 13 | 2142 | 71 | ||
| AK172168.1 | - | 12 - 13 | 1976 | 71 | ||
| BC004663.1 | - | 6 - 7 | 1028 | 71 | ||
| BC057867.1 | - | 12 - 13 | 1921 | 5327748 | 71 | |
| L33779.1 | - | 12 - 13 | 1940 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dsc2
Genomic Map