Assay Details
| Gene Symbol: Dsc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013505.3 | NP_038533.1 | 8 - 9 | 1328 | 87 | |
| GenBank mRNA | AK028604.1 | - | 9 - 10 | 1368 | 87 | |
| AK028636.1 | - | 8 - 9 | 1329 | 87 | ||
| AK030333.1 | - | 8 - 9 | 1329 | 87 | ||
| AK048547.1 | - | 8 - 9 | 1329 | 87 | ||
| AK172168.1 | - | 8 - 9 | 1163 | 87 | ||
| BC004663.1 | - | 2 - 3 | 215 | 87 | ||
| BC057867.1 | - | 8 - 9 | 1108 | 5327748 | 87 | |
| L33779.1 | - | 8 - 9 | 1127 | 87 | ||
| X73885.1 | - | 1 - 2 | 46 | 87 | ||
More Information
Set Membership: |
|

Dsc2
Genomic Map