Assay Details
| Gene Symbol: Dnajc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029669.4 | NP_083945.1 | 3 - 4 | 453 | 101 | |
| GenBank mRNA | AK012521.1 | - | 3 - 4 | 406 | 101 | |
| AK030483.1 | - | 3 - 4 | 431 | 101 | ||
| BC026452.1 | - | 3 - 4 | 415 | 4460764 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnajc18
Genomic Map