Assay Details
| Gene Symbol: Dsg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007883.2 | NP_031909.2 | 1 - 2 | 229 | 107 | |
| GenBank mRNA | AB072269.1 | - | 1 - 2 | 257 | 107 | |
| BC131653.1 | - | 1 - 2 | 205 | 40110529 | 107 | |
| BC131654.1 | - | 1 - 2 | 205 | 40110531 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB072269.1, BC131653.1, BC131654.1, NM_007883.2 transcripts.
Set Membership: |
|

Dsg2
Genomic Map