Assay Details
| Gene Symbol: Dsg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007883.2 | NP_031909.2 | 4 - 5 | 564 | 62 | |
| GenBank mRNA | AB072269.1 | - | 4 - 5 | 592 | 62 | |
| BC020144.1 | - | 4 - 5 | 458 | 62 | ||
| BC044848.1 | - | 4 - 5 | 525 | 62 | ||
| BC131653.1 | - | 4 - 5 | 540 | 40110529 | 62 | |
| BC131654.1 | - | 4 - 5 | 540 | 40110531 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dsg2
Genomic Map