| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600001469.mm.1 |
NM_007594.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001469.mm.1 |
NM_184053.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001469.mm.1 |
TC0600000229.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600001466.mm.1 |
NM_001285412.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001466.mm.1 |
NM_007594.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001466.mm.1 |
NM_184053.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001466.mm.1 |
TC0600000229.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600001470.mm.1 |
NM_001285412.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001470.mm.1 |
NM_007594.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001470.mm.1 |
NM_184053.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001470.mm.1 |
TC0600000229.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600001467.mm.1 |
NM_001285412.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001467.mm.1 |
NM_007594.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001467.mm.1 |
NM_184053.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001467.mm.1 |
TC0600000229.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600001475.mm.1 |
NM_184053.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600001475.mm.1 |
TC0600000229.mm.1 |
MTA TC |