Assay Details
| Gene Symbol: Ywhaz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011740.3 | NP_035870.1 | 1 - 2 | 396 | 56 | |
| GenBank mRNA | BB652039.1 | - | 1 - 2 | 396 | 56 | |
| BC050891.1 | - | 1 - 2 | 102 | 6419584 | 56 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC050891.1, NM_011740.3 transcripts.
Set Membership: |
|

Ywhaz
Genomic Map