Assay Details
| Gene Symbol: Ndufa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010885.5 | NP_035015.2 | 2 - 3 | 275 | 74 | |
| GenBank mRNA | AF124786.1 | - | 2 - 3 | 227 | 74 | |
| AK003608.1 | - | 2 - 3 | 237 | 74 | ||
| AK018732.1 | - | 2 - 3 | 213 | 74 | ||
| AV482131.1 | - | 2 - 3 | 275 | 74 | ||
| BC006815.1 | - | 2 - 3 | 213 | 3155763 | 74 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31625586 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31625589 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31625584 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31625587 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31625590 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384946 | AK003608.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384946 | AK018732.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384946 | BC006815.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384946 | NM_010885.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384946 | AF124786.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384946 | AK003608.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384946 | AK018732.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384946 | BC006815.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384946 | NM_010885.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P384946 | AF124786.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 243156 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 67964 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103273 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384946 | AK003608.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384946 | AK018732.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384946 | BC006815.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384946 | NM_010885.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384946 | AF124786.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384946 | AK003608.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384946 | AK018732.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384946 | BC006815.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384946 | NM_010885.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P384946 | AF124786.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384946 | AK003608.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384946 | AK018732.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384946 | BC006815.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384946 | NM_010885.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384946 | AF124786.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384946 | AK003608.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384946 | AK018732.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384946 | BC006815.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384946 | NM_010885.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P384946 | AF124786.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625576 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625589 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625574 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625581 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625583 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625586 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625587 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625584 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625575 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625570 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31625578 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103273 | - | - |
| Whole Mouse Genome G4122A | A_51_P384946 | AK003608.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384946 | AK018732.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384946 | BC006815.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384946 | NM_010885.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384946 | AF124786.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384946 | AK003608.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384946 | AK018732.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384946 | BC006815.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384946 | NM_010885.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P384946 | AF124786.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128413 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115942 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103273 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13924926 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14050352 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14050343 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14050349 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14050346 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31625587 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31625589 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31625586 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31625590 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31625584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625583 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625587 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31625574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625580 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625583 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625574 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625578 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625570 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625576 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625589 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31625587 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10458387 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10458388 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10458389 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10458390 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417368_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10458386 | - | - |
| Mouse Genome 430 2.0 Array | 1417368_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1417368_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17991_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17991_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1800008844.mm.1 | NM_010885.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800008844.mm.1 | TC1800001198.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800008845.mm.1 | NM_010885.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800008845.mm.1 | TC1800001198.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417368_PM_s_at | - | - |
| Mouse Expression Set 430 | 1417368_s_at | - | - |
| Murine Genome U74v2 Set | 96280_at | - | - |
| Murine Genome U74v2 Set | 96280_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417368_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417368_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417368_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417368_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6020100 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006020100 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2588601 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2588601 | - | - |
| mouse-6 v1.1 (Illumina) | scl017991.2_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6020100 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl017991.2_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6020100 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2588601 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2588601 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl017991.2_1-S | - | - |

Ndufa2
Genomic Map