Assay Details
| Gene Symbol: Pcdha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054072.1 | NP_473413.1 | 1 | 2337 | 62 | |
| GenBank mRNA | AY013756.1 | - | 1 | 2337 | 62 | |
| BC082326.1 | - | 1 | 2438 | 30356350 | 62 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pcdha1
Genomic Map