| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800001740.mm.1 |
NM_001170966.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001740.mm.1 |
NM_001170966.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001740.mm.1 |
NM_001170967.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001740.mm.1 |
NM_001170967.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001740.mm.1 |
NM_001170970.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001740.mm.1 |
NM_001170970.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001740.mm.1 |
NM_028866.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001740.mm.1 |
NM_028866.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001740.mm.1 |
TC1800000244.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800001739.mm.1 |
NM_001170966.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001739.mm.1 |
NM_001170966.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001739.mm.1 |
NM_001170967.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001739.mm.1 |
NM_001170967.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001739.mm.1 |
NM_001170970.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001739.mm.1 |
NM_001170970.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001739.mm.1 |
NM_028866.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001739.mm.1 |
NM_028866.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001739.mm.1 |
TC1800000244.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800001742.mm.1 |
NM_001170966.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001742.mm.1 |
NM_001170966.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001742.mm.1 |
NM_001170967.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001742.mm.1 |
NM_001170967.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001742.mm.1 |
NM_001170970.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001742.mm.1 |
NM_001170970.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001742.mm.1 |
NM_028866.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001742.mm.1 |
NM_028866.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800001742.mm.1 |
TC1800000244.mm.1 |
MTA TC |