Assay Details
| Gene Symbol: Erbb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010153.1 | NP_034283.1 | 9 - 10 | 1107 | 64 | |
| GenBank mRNA | AY686636.1 | - | 9 - 10 | 1107 | 64 | |
| BC106091.1 | - | 1258 | 30844039 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Erbb3
Genomic Map