Assay Details
| Gene Symbol: Csn1s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286015.1 | NP_001272944.1 | 17 - 18 | 539 | 72 | |
| NM_001286016.1 | NP_001272945.1 | 18 - 19 | 563 | 72 | ||
| NM_007784.3 | NP_031810.1 | 18 - 19 | 563 | 72 | ||
| GenBank mRNA | BC002101.1 | - | 18 - 19 | 532 | 3484624 | 72 |
| BC003859.1 | - | 18 - 19 | 547 | 3486384 | 72 | |
| BC006024.1 | - | 17 - 18 | 523 | 3592625 | 72 | |
| BC007192.1 | - | 18 - 19 | 547 | 72 | ||
| BC008278.1 | - | 18 - 19 | 547 | 2812523 | 72 | |
| BC040246.1 | - | 18 - 19 | 559 | 2647887 | 72 | |
| M36780.1 | - | 17 - 18 | 502 | 72 | ||
More Information
Set Membership: |
|

Csn1s1
Genomic Map