Assay Details
| Gene Symbol: Csn1s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286015.1 | NP_001272944.1 | 22 - 23 | 625 | 65 | |
| NM_001286016.1 | NP_001272945.1 | 23 - 24 | 649 | 65 | ||
| NM_007784.3 | NP_031810.1 | 23 - 24 | 649 | 65 | ||
| GenBank mRNA | AK085690.1 | - | 23 - 24 | 653 | 65 | |
| BC002101.1 | - | 23 - 24 | 618 | 3484624 | 65 | |
| BC003859.1 | - | 23 - 24 | 633 | 3486384 | 65 | |
| BC006024.1 | - | 22 - 23 | 609 | 3592625 | 65 | |
| BC007192.1 | - | 23 - 24 | 633 | 65 | ||
| BC008278.1 | - | 23 - 24 | 633 | 2812523 | 65 | |
| BC020321.1 | - | 4 - 5 | 103 | 65 | ||
| BC040246.1 | - | 23 - 24 | 645 | 2647887 | 65 | |
| M36780.1 | - | 22 - 23 | 588 | 65 | ||
More Information
Set Membership: |
|

Csn1s1
Genomic Map