Assay Details
| Gene Symbol: Csn1s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286015.1 | NP_001272944.1 | 24 - 25 | 667 | 63 | |
| NM_001286016.1 | NP_001272945.1 | 25 - 26 | 691 | 63 | ||
| NM_007784.3 | NP_031810.1 | 25 - 26 | 691 | 63 | ||
| GenBank mRNA | AK085690.1 | - | 25 - 26 | 695 | 63 | |
| BC002101.1 | - | 25 - 26 | 660 | 3484624 | 63 | |
| BC003859.1 | - | 25 - 26 | 675 | 3486384 | 63 | |
| BC006024.1 | - | 24 - 25 | 651 | 3592625 | 63 | |
| BC007192.1 | - | 25 - 26 | 675 | 63 | ||
| BC008278.1 | - | 25 - 26 | 675 | 2812523 | 63 | |
| BC020321.1 | - | 6 - 7 | 145 | 63 | ||
| BC040246.1 | - | 25 - 26 | 687 | 2647887 | 63 | |
| M36780.1 | - | 24 - 25 | 630 | 63 | ||
More Information
Set Membership: |
|

Csn1s1
Genomic Map