Assay Details
| Gene Symbol: Csn1s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286015.1 | NP_001272944.1 | 26 - 27 | 699 | 94 | |
| NM_001286016.1 | NP_001272945.1 | 27 - 28 | 723 | 94 | ||
| NM_007784.3 | NP_031810.1 | 27 - 28 | 723 | 94 | ||
| GenBank mRNA | AK085690.1 | - | 27 - 28 | 727 | 94 | |
| BC002101.1 | - | 27 - 28 | 692 | 3484624 | 94 | |
| BC003859.1 | - | 27 - 28 | 707 | 3486384 | 94 | |
| BC006024.1 | - | 26 - 27 | 683 | 3592625 | 94 | |
| BC007192.1 | - | 27 - 28 | 707 | 94 | ||
| BC008278.1 | - | 27 - 28 | 707 | 2812523 | 94 | |
| BC020321.1 | - | 8 - 9 | 177 | 94 | ||
| BC040246.1 | - | 27 - 28 | 719 | 2647887 | 94 | |
| M36780.1 | - | 26 - 27 | 662 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Csn1s1
Genomic Map