Assay Details
| Gene Symbol: Zyx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289617.1 | NP_001276546.1 | 3 - 4 | 711 | 148 | |
| NM_011777.3 | NP_035907.1 | 3 - 4 | 711 | 148 | ||
| GenBank mRNA | AK031415.1 | - | 3 - 4 | 509 | 148 | |
| AK035743.1 | - | 3 - 4 | 482 | 148 | ||
| AK076491.1 | - | 3 - 4 | 482 | 148 | ||
| AK090110.1 | - | 3 - 4 | 478 | 148 | ||
| AK147812.1 | - | 3 - 4 | 712 | 148 | ||
| AK170571.1 | - | 3 - 4 | 485 | 148 | ||
| AK205275.1 | - | 2 - 3 | 290 | 148 | ||
| X99063.1 | - | 3 - 4 | 669 | 148 | ||
| Y07711.1 | - | 2 - 3 | 419 | 148 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zyx
Genomic Map