Assay Details
| Gene Symbol: Zyx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289617.1 | NP_001276546.1 | 8 - 9 | 1773 | 61 | |
| NM_001289618.1 | NP_001276547.1 | 7 - 8 | 1680 | 61 | ||
| NM_001289619.1 | NP_001276548.1 | 7 - 8 | 1680 | 61 | ||
| NM_011777.3 | NP_035907.1 | 8 - 9 | 1773 | 61 | ||
| GenBank mRNA | AK031415.1 | - | 8 - 9 | 1571 | 61 | |
| AK035743.1 | - | 8 - 9 | 1544 | 61 | ||
| AK076491.1 | - | 8 - 9 | 1544 | 61 | ||
| AK090110.1 | - | 8 - 9 | 1540 | 61 | ||
| AK147812.1 | - | 8 - 9 | 1774 | 61 | ||
| AK161709.1 | - | 7 - 8 | 1477 | 61 | ||
| AK170571.1 | - | 8 - 9 | 1547 | 61 | ||
| BC054775.1 | - | 7 - 8 | 1639 | 6808149 | 61 | |
| Y07711.1 | - | 7 - 8 | 1481 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zyx
Genomic Map