Assay Details
| Gene Symbol: Tmem106b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027992.3 | NP_082268.2 | 6 - 7 | 891 | 91 | |
| GenBank mRNA | AK009646.1 | - | 6 - 7 | 794 | 91 | |
| AK148477.1 | - | 6 - 7 | 815 | 91 | ||
| BC050246.1 | - | 6 - 7 | 808 | 6313498 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem106b
Genomic Map