Assay Details
| Gene Symbol: Mtdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026002.4 | NP_080278.3 | 3 - 4 | 881 | 73 | |
| GenBank mRNA | AF501309.1 | - | 3 - 4 | 819 | 73 | |
| AK029915.1 | - | 3 - 4 | 882 | 73 | ||
| AK151269.1 | - | 3 - 4 | 865 | 73 | ||
| AK151810.1 | - | 3 - 4 | 876 | 73 | ||
| BC023115.1 | - | 3 - 4 | 780 | 73 | ||
| BC036994.1 | - | 3 - 4 | 600 | 73 | ||
| BC138858.1 | - | 3 - 4 | 595 | 8861880 | 73 | |
| BC138859.1 | - | 3 - 4 | 595 | 8861881 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtdh
Genomic Map