Assay Details
| Gene Symbol: Mtdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026002.4 | NP_080278.3 | 5 - 6 | 1127 | 143 | |
| GenBank mRNA | AF501309.1 | - | 5 - 6 | 1065 | 143 | |
| AK151269.1 | - | 5 - 6 | 1111 | 143 | ||
| AK151810.1 | - | 5 - 6 | 1122 | 143 | ||
| AY553638.1 | - | 5 - 6 | 810 | 143 | ||
| BC023115.1 | - | 5 - 6 | 1026 | 143 | ||
| BC036994.1 | - | 5 - 6 | 846 | 143 | ||
| BC138858.1 | - | 5 - 6 | 841 | 8861880 | 143 | |
| BC138859.1 | - | 5 - 6 | 841 | 8861881 | 143 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mtdh
Genomic Map