Assay Details
| Gene Symbol: Dlx6os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015388.1 | 1 - 2 | 225 | 70 | ||
| GenBank mRNA | AK032537.1 | - | 1 - 2 | 225 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | split hand/foot malformation (ectrodactyly) type 1 | Shfm1 |
DSS1, Shfdg1, Shfg |
-245.3kb | Sense | Chr. 6 - 6558275 - 6578658 on NCBI Build GRCm38 |
| Downstream | distal-less homeobox 5 | Dlx5 | AI385752 | 8.3kb | Sense | Chr. 6 - 6877801 - 6882068 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P112463 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 322805 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 353630 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 70733 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336589 | AK047127.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336589 | AK077101.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336589 | AK077503.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336589 | AK170524.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336589 | AK171511.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336589 | BC025632.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336589 | BC026926.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336589 | BC031913.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336589 | NM_146170.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61319 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 393128 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 373471 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 615399 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 31976 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 451744 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 103006 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 142591 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 218086 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 758032 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 770420 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 924515 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546607 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931856 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 952062 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 331877 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 612120 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 764428 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 972 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 785589 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 291960 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 500453 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 656188 | AK046793.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 656188 | AK161890.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 656188 | BC049167.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 656188 | BC070410.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 656188 | NM_175285.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 63589 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 157262 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 173709 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 102900 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 79408 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 43993 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P163938 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165076 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P112463 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05802579 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165076 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P163938 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05802528 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05802487 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05802513 | - | - |
| Whole Mouse Genome G4122A | A_52_P231012 | NR_015388.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P231012 | AK032180.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P231012 | AK032537.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P231012 | AK038694.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P231012 | AK044034.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P112463 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134126 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P163938 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165076 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05802487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218489 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218523 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05398346 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05361983 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05471675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218620 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218541 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218567 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05779584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05268272 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218575 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218618 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05243806 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05550539 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218608 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05051730 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05105583 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218526 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218571 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218490 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218637 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218485 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05471667 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218505 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218476 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218474 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218519 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05871461 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05764123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218555 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05100565 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05871466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05871463 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218630 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218613 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05779576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05434843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218581 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05105584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218471 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218547 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05779570 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218530 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218550 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218565 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218556 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218538 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218544 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218596 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05471672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05126171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05871451 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218564 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218622 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218475 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218602 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218525 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218511 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218619 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05550540 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218542 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218540 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218516 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218518 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218634 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218605 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218521 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218560 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05779592 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05243796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218484 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P05361984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24218606 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1438799_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438799_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438799_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600014446.mm.1 | NR_015388.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600014446.mm.1 | NR_015388.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600014446.mm.1 | TC0600001830.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600014445.mm.1 | NR_015388.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600014445.mm.1 | NR_015388.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600014445.mm.1 | TC0600001830.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600014443.mm.1 | NR_015388.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600014443.mm.1 | NR_015388.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600014443.mm.1 | TC0600001830.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600014444.mm.1 | NR_015388.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600014444.mm.1 | NR_015388.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600014444.mm.1 | TC0600001830.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600014454.mm.1 | NR_015388.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600014454.mm.1 | NR_015388.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600014454.mm.1 | TC0600001830.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1438799_PM_at | - | - |
| Mouse Expression Set 430 | 1438799_at | - | - |
| Murine Genome U74v2 Set | 138093_PM_f_at | - | - |
| Murine Genome U74v2 Set | 138093_f_at | - | - |
| Murine Genome U74v2 Set | 170818_PM_i_at | - | - |
| Murine Genome U74v2 Set | 170818_i_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438799_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438799_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 668336 | - | - |

Dlx6os1
Genomic Map