Assay Details
| Gene Symbol: Abca14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026458.4 | NP_080734.3 | 12 - 13 | 160 | ||
| GenBank mRNA | AK029665.1 | - | 13 - 14 | 160 | ||
| AY243470.1 | - | 12 - 13 | 1863 | 160 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abca14
Genomic Map