Assay Details
Gene Symbol: Abca14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026458.4 | NP_080734.3 | 13 - 14 | 2024 | 112 | |
GenBank mRNA | AK029665.1 | - | 14 - 15 | 2136 | 112 | |
AY243470.1 | - | 13 - 14 | 2024 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
