Assay Details
| Gene Symbol: Abca14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026458.4 | NP_080734.3 | 27 - 28 | 4633 | 101 | |
| GenBank mRNA | AY243470.1 | - | 27 - 28 | 4633 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abca14
Genomic Map