Assay Details
Gene Symbol: Abca14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026458.4 | NP_080734.3 | 10 - 11 | 1609 | 55 | |
GenBank mRNA | AK029665.1 | - | 11 - 12 | 1721 | 55 | |
AY243470.1 | - | 10 - 11 | 1609 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
