Assay Details
| Gene Symbol: Lyrm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029610.3 | NP_083886.1 | 1 - 2 | 278 | 119 | |
| GenBank mRNA | AK009135.1 | - | 1 - 2 | 281 | 119 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK009135.1, NM_029610.3 transcripts.
Set Membership: |
|

Lyrm1
Genomic Map