Assay Details
| Gene Symbol: Cntnap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004357.2 | NP_001004357.2 | 19 - 20 | 3481 | 61 | |
| GenBank mRNA | AK129232.1 | - | 19 - 20 | 3454 | 61 | |
| BC064467.1 | - | 14 - 15 | 2372 | 61 | ||
More Information
Set Membership: |
|

Cntnap2
Genomic Map