| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500010971.mm.1 |
NM_001102458.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500010971.mm.1 |
NM_001102458.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500010971.mm.1 |
NM_018745.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500010971.mm.1 |
NM_018745.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500010971.mm.1 |
TC1500001412.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500010975.mm.1 |
NM_001102458.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500010975.mm.1 |
NM_001102458.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500010975.mm.1 |
NM_018745.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500010975.mm.1 |
NM_018745.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500010975.mm.1 |
TC1500001412.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500010969.mm.1 |
NM_001102458.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500010969.mm.1 |
NM_001102458.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500010969.mm.1 |
TC1500001412.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500010976.mm.1 |
NM_001102458.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500010976.mm.1 |
NM_001102458.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500010976.mm.1 |
NM_018745.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500010976.mm.1 |
NM_018745.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500010976.mm.1 |
TC1500001412.mm.1 |
MTA TC |