Assay Details
| Gene Symbol: Azin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102458.1 | NP_001095928.1 | 5 - 6 | 1033 | 68 | |
| NM_001301688.1 | NP_001288617.1 | 4 - 5 | 873 | 68 | ||
| NM_018745.5 | NP_061215.1 | 4 - 5 | 873 | 68 | ||
| NR_125913.1 | 4 - 5 | 873 | 68 | |||
| GenBank mRNA | AF032128.1 | - | 2 - 3 | 299 | 68 | |
| AK049680.1 | - | 4 - 5 | 874 | 68 | ||
| AK088112.1 | - | 4 - 5 | 871 | 68 | ||
| AK088671.1 | - | 5 - 6 | 1030 | 68 | ||
| AK133914.1 | - | 4 - 5 | 869 | 68 | ||
| BC019412.1 | - | 4 - 5 | 861 | 5151406 | 68 | |
| BC021152.1 | - | 4 - 5 | 826 | 68 | ||
| BC043722.1 | - | 2 - 3 | 379 | 5321733 | 68 | |
| BC046814.1 | - | 4 - 5 | 776 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Azin1
Genomic Map