Assay Details
| Gene Symbol: Azin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102458.1 | NP_001095928.1 | 6 - 7 | 1197 | 154 | |
| NM_001301688.1 | NP_001288617.1 | 5 - 6 | 1037 | 154 | ||
| NM_018745.5 | NP_061215.1 | 5 - 6 | 1037 | 154 | ||
| GenBank mRNA | AF032128.1 | - | 3 - 4 | 463 | 154 | |
| AK049680.1 | - | 5 - 6 | 1038 | 154 | ||
| AK088671.1 | - | 6 - 7 | 1194 | 154 | ||
| AK133914.1 | - | 5 - 6 | 1033 | 154 | ||
| BC019412.1 | - | 5 - 6 | 1025 | 5151406 | 154 | |
| BC021152.1 | - | 5 - 6 | 990 | 154 | ||
| BC043722.1 | - | 3 - 4 | 543 | 5321733 | 154 | |
More Information
Updated Mapping Information
Set Membership: |
|

Azin1
Genomic Map