Assay Details
| Gene Symbol: Azin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102458.1 | NP_001095928.1 | 8 - 9 | 1419 | 74 | |
| NM_001301688.1 | NP_001288617.1 | 7 - 8 | 1259 | 74 | ||
| NM_018745.5 | NP_061215.1 | 7 - 8 | 1259 | 74 | ||
| NR_125913.1 | 7 - 8 | 1411 | 74 | |||
| GenBank mRNA | AF032128.1 | - | 5 - 6 | 685 | 74 | |
| AK049680.1 | - | 7 - 8 | 1260 | 74 | ||
| AK088112.1 | - | 7 - 8 | 1257 | 74 | ||
| AK088671.1 | - | 8 - 9 | 1416 | 74 | ||
| AK133914.1 | - | 7 - 8 | 1255 | 74 | ||
| BC019412.1 | - | 7 - 8 | 1247 | 5151406 | 74 | |
| BC021152.1 | - | 7 - 8 | 1212 | 74 | ||
| BC043722.1 | - | 5 - 6 | 765 | 5321733 | 74 | |
| BC046814.1 | - | 7 - 8 | 1314 | 74 | ||
| CF738062.1 | - | 3 - 4 | 250 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Azin1
Genomic Map